Anti-ATP5H Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # USBI032285-FITC
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:ATP synthase, H+ transporting, mitochondrial Fo complex, subunit d
- Antigen symbol:ATP5H
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Form:liquid
- Amino acid number:75 to 105
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: ATP5H (ATP synthase, H+ transporting, mitochondrial Fo complex, subunit d)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human