Order Entry
Sweden
ContactUsLinkComponent
Anti-APOBEC3B Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anti-APOBEC3B Rabbit Polyclonal Antibody (APC (Allophycocyanin))
  USBI031980-APC
undefined
Anti-APOBEC3B Rabbit Polyclonal Antibody (APC (Allophycocyanin))
  USBI031980-APC

 

  • Antibody type:
    Primary
  • Antigen name:
    Apolipoprotein B mRNA editing enzyme, catalytic polypeptide 3B
  • Antigen symbol:
    APOBEC3B
  • Clonality:
    Polyclonal
  • Conjugation:
    APC (Allophycocyanin)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Form:
    liquid
  • Amino acid number:
    307 to 337
  • Storage buffer:
    PBS, pH 7,2, contains ~0,09% sodium azide
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    200 µl
  • Pk:
    200 µl

 

 

Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-100
Do not freeze!

Type: Primary
Antigen: APOBEC3B (apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3B)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse