Anti-Myc Tag Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 9E10]
Catalog # USBI031143-HRP
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Myc tag
- Antigen symbol:MYC
- Clonality:Monoclonal
- Clone:9E10
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:3000; Western Blot: 1:2000
Type: Primary
Antigen: Myc Tag (Myc tag)
Clonality: Monoclonal
Clone: 90000000000
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: