Anti-HIPK4 Mouse Monoclonal Antibody [clone: 292CT1.4.1]
Catalog # USBI031016
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:homeodomain interacting Protein kinase 4
- Antigen symbol:HIPK4
- Clonality:Monoclonal
- Clone:292CT1.4.1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgM
- Reactivity:Human
- Western blot:Yes
- Format:ascites
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:500-8000
Type: Primary
Antigen: HIPK4 (homeodomain interacting protein kinase 4)
Clonality: Monoclonal
Clone: 292CT1.4.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human