Order Entry
Sweden
ContactUsLinkComponent
Anti-CD34 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 132CT29.1.5]
Anti-CD34 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 132CT29.1.5]
  USBI030848-FITC
undefined
Anti-CD34 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 132CT29.1.5]
  USBI030848-FITC

 

  • Antibody type:
    Primary
  • Antigen name:
    CD34 molecule
  • Antigen symbol:
    CD34
  • Clonality:
    Monoclonal
  • Clone:
    132CT29.1.5
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    internal
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, contains ~0,09% sodium azide
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µl
  • Pk:
    200 µl

 

 

Recommended dilution: FLISA: 1:1000; Western Blot: 1:500-1000; Immunochemistry: 1:50-100

Type: Primary
Antigen: CD34 (CD34 molecule)
Clonality: Monoclonal
Clone: 132CT29.1.5
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse