Anti-Neuronal Marker, Pan Rabbit Polyclonal Antibody (Cy3®)
Catalog # USBI029559
Leverantör: US Biological
Specifikationer
- Antibody type:Primary
- Antigen name:Neuronal Marker, Pan
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Rat,Mouse
- Form:liquid
- Storage buffer:PBS, 15mg/ml BSA, 0,05% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 µl
- Pk:100 µl
Specifikationer
Om denna produkt
Type: Primary
Antigen: Neuronal Marker, Pan
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat