Order Entry
Sweden
ContactUsLinkComponent
Anti-Neuronal Marker, Pan Rabbit Polyclonal Antibody
Anti-Neuronal Marker, Pan Rabbit Polyclonal Antibody
  USBI029557
Anti-Neuronal Marker, Pan Rabbit Polyclonal Antibody
  USBI029557

 

  • Antibody type:
    Primary
  • Antigen name:
    Neuronal Marker, Pan
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Rat,
    Mouse
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,05% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Type: Primary
Antigen: Neuronal Marker, Pan
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat