Anti-Bystin Rabbit Polyclonal Antibody
Catalog # USBI029429
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Bystin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Form:liquid
- Storage buffer:0,1M Tris-glycine, pH 7,4, 150mM sodium chloride, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1 ?g/ml; Immunochemistry: 1:300
Type: Primary
Antigen: Bystin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse