Order Entry
Sweden
ContactUsLinkComponent
 
Anti-ADGRE1 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: BM8]
Catalog # ROCK200-508-L55
undefined
Anti-ADGRE1 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: BM8]
Catalog # ROCK200-508-L55

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    adhesion G protein-coupled receptor E1
  • Antigen symbol:
    ADGRE1
  • Clonality:
    Monoclonal
  • Clone:
    BM8
  • Conjugation:
    RPE (R-Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Mouse
  • Cross adsorption:
    none
  • Format:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    13733
  • Antigen synonyms:
    Ly71|Gpf480|F4/80|TM7LN3|DD7A5-7|EGF-TM7|Emr1
  • Storage buffer:
    0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide,
  • Storage temperature:
    Cool
  • Concentration:
    0,2 mg/ml Sufficient to run approximately 100 tests
  • Shipping temperature:
    Ice
  • Immunogen:
    Anti-F4/80 Antibody (Monoclonal) was produced by repeated immunizations with murine macrophages.
  • Tested applications:
    Fluoresence ELISA
  • Purification:
    affinity-purified
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Anti-F4/80 is useful for Flow Cytometry and immunochemistry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze! Light sensitive!

Type: Primary
Antigen: ADGRE1
Clonality: Monoclonal
Clone: BM8
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse