Anti-ADGRE1 Rat Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: BM8.1]
ROCK200-502-L55
- Antibody type:Primary
- Antigen name:adhesion G protein-coupled receptor E1
- Antigen symbol:ADGRE1
- Clonality:Monoclonal
- Clone:BM8.1
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:13733
- Antigen synonyms:Ly71|Gpf480|F4/80|TM7LN3|DD7A5-7|EGF-TM7|Emr1
- Storage buffer:0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, 0,1% Gelatin
- Storage temperature:Cool
- Concentration:0,5 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:Anti-F4/80 Antibody (Monoclonal) was produced by repeated immunizations with F4/80 antigen.
- Tested applications:Fluoresence ELISA
- Purification:affinity-purified
- Size:500 µg
- Pk:500 µG
Anti-F4/80 is useful for Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.
Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg); Sufficient to run approximately 500 tests
Do not freeze!
Type: Primary
Antigen: ADGRE1
Clonality: Monoclonal
Clone: BM8.1
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse