Order Entry
Sweden
ContactUsLinkComponent
Anti-IgG (H+L) Rabbit Polyclonal Antibody
Anti-IgG (H+L) Rabbit Polyclonal Antibody
Catalog # ROCK605-4102
undefined
Anti-IgG (H+L) Rabbit Polyclonal Antibody
Catalog # ROCK605-4102

Specifications

  • Antikroppstyp:
    Sekundär
  • Antigensymbol:
    IgG (H+L)
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flödescytometri:
    Yes
  • Gelskifte:
    Yes
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • ImmunoFluorescens:
    Yes
  • Immunutfällning:
    Yes
  • Reaktivitet:
    Goat
  • Western blot:
    Yes
  • Korsad adsorption:
    none
  • ChIP:
    Yes
  • Neutralisation:
    Yes
  • Mått:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
  • Förvaringstemperatur:
    Cool
  • Koncentration:
    2,0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Immunogen:
    Goat IgG whole molecule
  • Rening:
    purified by immunoaffinity chromatography
  • Storlek:
    2 mg
  • Förp.:
    2 mg

Specifications

About this item

Secondary antibody reagents are ideal for western blotting, immunochemistry, ELISA, Fluorescence Microscopy, Flow Cytometry as well as other antibody detection methods.

Recommended Dilutions: ELISA: 1:800,000; Western Blot: 1:10000 - 1:60,000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Goat