Anti-HP Rabbit Polyclonal Antibody
ROCK600-401-272
- Antibody type:Primary
- Antigen name:Haptoglobin
- Antigen symbol:HP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:3240
- Antigen synonyms:HPA1S|BP|HP2ALPHA2
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:2,5 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:Human Haptoglobin Antibody was produced by repeated immunizations with Human Haptoglobin Protein.
- Purification:purified by immunoaffinity chromatography
- Size:1 mg
- Pk:1 mg
Human Haptoglobin Antibody is suitable for western blotting, ELISA and IHC. Researchers should determine optimal titers for applications that are not stated below.
Recommended Dilutions: ELISA: 1:500,000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Type: Primary
Antigen: HP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human