Order Entry
Sweden
ContactUsLinkComponent
Anti-GST Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Anti-GST Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # ROCK600-105-200
undefined
Anti-GST Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # ROCK600-105-200

Specifications

  • Antibody type:
    Tag
  • Antigen name:
    Glutathione-S-Transferase
  • Antigen symbol:
    GST
  • Clonality:
    Polyclonal
  • Conjugation:
    AP (Alkaline Phosphatase)
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Multiple
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    1 mg
  • Pk:
    1 mg

Specifications

About this item

Type: Primary
Antigen: GST
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype:
Reactivity: Pan