Anti-RPE Goat Polyclonal Antibody
ROCK600-101-387
- Antibody type:Tag
- Antigen symbol:R-Phycoerythrin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Cat
- Western blot:Yes
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:1,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:This affinity purified antibody was prepared from whole goat serum produced by repeated immunizations with highly purified R-Phycoerythrin from the seaweed gracilaria.
- Purification:purified by immunoaffinity chromatography
- Size:1 mg
- Pk:1 mg
Suitable for ELISA, western blotting, fluorescent assays, immunoprecipitation, immunodiffusion, conjugation to magnetic particles (beads) and most other immunological methods requiring lot-to-lot consistency, high titer and specificity.
Recommended Dilutions: ELISA: 1:20000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Type: Tag
Antigen: RPE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: