Anti-APOE Goat Polyclonal Antibody
ROCK600-101-197
- Antibody type:Primary
- Antigen name:apolipoprotein E
- Antigen symbol:APOE
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:348
- Antigen synonyms:APO-E|AD2|LDLCQ5|LPG
- Storage buffer:0,125 M Sodium Borate, 0,075 M Sodium Chloride, 0,005 M EDTA, pH 8.0, 0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:1,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:ApolipoProtein Type E was isolated from human plasma by density gradient centrifugation followed by HPLC purification.
- Purification:purified by immunoaffinity chromatography
- Size:500 µg
- Pk:500 µG
ApolipoProtein E antibodies have been used for indirect trapping ELISA for quantitation of antigen in serum using a standard curve, for immunoprecipitation and for western blotting for highly sensitive qualitative analysis.
Recommended Dilutions: ELISA: 1:4000 - 1:8000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:50 - 1:200; Immunoprecipitation: 1:100;
Type: Primary
Antigen: APOE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Rat