Order Entry
Sweden
ContactUsLinkComponent
Anti-IgG (H+L) Goat Polyclonal Antibody
Anti-IgG (H+L) Goat Polyclonal Antibody
  ROCK219-101-002
Anti-IgG (H+L) Goat Polyclonal Antibody
  ROCK219-101-002

 

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG (H+L)
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Armenian hamster
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide
  • Storage temperature:
    Cool
  • Concentration:
    10,0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Size:
    50 mg
  • Pk:
    50 mg

 

 

Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host:
Isotype:
Reactivity: Armenian Hamster