Order Entry
Sweden
ContactUsLinkComponent
 
Anti-IgG F(ab')2 Rabbit Polyclonal Antibody
  ROCK110-4104
Anti-IgG F(ab')2 Rabbit Polyclonal Antibody
  ROCK110-4104

 

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG F(ab')2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Gel shift:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • ChIP:
    Yes
  • Neutralisation:
    Yes
  • Format:
    Antiserum
  • Form:
    Lyophilized
  • Storage buffer:
    0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2,
  • Storage temperature:
    Cool
  • Concentration:
    28,0 mg/ml by Refractometry
  • Shipping temperature:
    Ice
  • Immunogen:
    Mouse IgG F(ab')2 fragment
  • Purification:
    delipidation and defibrination
  • Size:
    2 ml
  • Pk:
    2 mL

 

 

Secondary Rabbit Anti-IgG F(ab')2 Reacts with Mouse

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(ab')2
Host: Rabbit
Isotype: IgG
Reactivity: Mouse