Anti-CCNE1 Rabbit Polyclonal Antibody
Catalog # ROCK100-401-156
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:cyclin E1
- Antigen symbol:CCNE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Format:Antiserum
- Form:Liquid (sterile filtered)
- Gene ID:898
- Antigen synonyms:pCCNE1|CCNE
- Storage buffer:0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:70 mg/ml by Refractometry
- Shipping temperature:Ice
- Immunogen:Anti-Cyclin-E Antibody was produced by repeated immunizations with a KLH conjugated peptide corresponding to a region near the carboxy terminus of human Cyclin E.
- Purification:delipidation and defibrination
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Anti-Cyclin-E antibody is suitable for the detection by immunoblot, ELISA and IP. Researchers should determine optimal titers for applications that are not stated below. Antiserum will specifically react with a 45 kDa Cyclin E Protein from human tissue.
Recommended Dilutions: ELISA: 1:100000; Western Blot: 1:2000 - 1:10000; Immunoprecipitation: 1:100;
Type: Primary
Antigen: CCNE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human