Anti-Raspberry Bushy Dwarf Virus Chicken Polyclonal Antibody
QEDB18701
- Antibody type:Primary
- Antigen name:Raspberry Bushy Dwarf Virus
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Chicken
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:250 µg
- Pk:250 µG
Type: Primary
Antigen: Raspberry Bushy Dwarf Virus
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: none
Reactivity: