Order Entry
Sweden
ContactUsLinkComponent
 
Anti-Neurofilament H (SMI-31) Mouse Monoclonal Antibody
  NE1022-100
Anti-Neurofilament H (SMI-31) Mouse Monoclonal Antibody
  NE1022-100

 

  • Antibody type:
    Primary
  • Antigen name:
    SMI-31
  • Antigen symbol:
    Neurofilament H
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Type: Primary
Antigen: Neurofilament H (SMI-31)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: