Anti-Ig Sheep Polyclonal Antibody (Biotin)
Catalog # RPN1003-2ML
Supplier: Cytiva
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin
- Antigen symbol:Ig
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Sheep
- ImmunoChemistry:Yes
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:2 ml
- Pk:2 ml
Specifications
About this item
Type: Secondary
Antigen: Ig
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: none
Reactivity: Human