Anti-BATF Goat polyclonal antibody unconjugated
Catalog # EVBIEB12083
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:basic leucine zipper transcription factor, ATF-like
- Antigen symbol:BATF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Epitope:C-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: BATF (basic leucine zipper transcription factor, ATF-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: