Anti-creatine kinase M-type Goat polyclonal antibody unconjugated
Catalog # EVBIEB11997
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:creatine Kinase M-type
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Amino acid number:258 to 270
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: creatine kinase M-type
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: