Anti-TNNI3 Goat Polyclonal Antibody
EVBIEB11958
- Antibody type:Primary
- Antigen name:troponin I type 3 (cardiac)
- Antigen symbol:TNNI3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Amino acid number:117 to 127
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: TNNI3 (troponin I type 3 (cardiac))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: