Anti-SKA3 Goat Polyclonal Antibody
Katalog # EVBIEB09603
Leverantör: Everest Biotech
Specifikationer
- Antibody type:Primary
- Antigen name:spindle and kinetochore associated complex subunit 3
- Antigen symbol:Spindle and kinetochore-associated protein 3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Dog,Rat,Mouse
- Epitope:internal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifikationer
Om denna produkt
Type: Primary
Antigen: SKA3 (spindle and kinetochore associated complex subunit 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Mouse, Rat