Anti-CXCL2 Goat Polyclonal Antibody
Catalog # EVBIEB09352
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:chemokine (C-X-C motif) ligand 2
- Antigen symbol:CXCL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: CXCL2 (chemokine (C-X-C motif) ligand 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human