Anti-SH2D3A / NSP1 Goat Polyclonal Antibody
Catalog # EVBIEB09216
Supplier: Everest Biotech
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:SH2 domain containing 3A
- Antigensymbol:SH2D3A
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Goat
- Isotyp:IgG
- Reaktivitet:Human
- Western blot:Yes
- Epitop:internal
- Korsad adsorption:none
- Antigen synonymer:NSP1
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Storlek:100 µg
- Förp.:0,1 mg
Specifications
About this item
Type: Primary
Antigen: SH2D3A / NSP1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human