Anti-ABCB10 Goat Polyclonal Antibody
EVBIEB08701
- Antibody type:Primary
- Antigen name:ATP Binding Cassette, sub-family B, member 10
- Antigen symbol:ABCB10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Epitope:C-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: ABCB10 (ATP-binding cassette, sub-family B (MDR/TAP), member 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat