Anti-PAPIN / PDZK3 Goat Polyclonal Antibody
Katalog # EVBIEB06236
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen symbol:PAPIN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Epitope:N-terminal
- Cross adsorption:none
- Antigen synonyms:PDZK3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: PAPIN / PDZK3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity: Human