Anti-NOD1 Goat Polyclonal Antibody
Catalog # EVBIEB05389
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:Nucleotide-binding oligomerization domain containing 1
- Antigen symbol:NOD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: NOD1 (nucleotide-binding oligomerization domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: