Anti-Paxillin Mouse Monoclonal Antibody [clone: PAX-14]
BSBTMA1080
- Antibody type:Primary
- Antigen name:Paxillin
- Antigen symbol:PXN
- Clonality:Monoclonal
- Clone:PAX-14
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Chicken,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Antigen synonyms:paxillin|PAX-1|paxillin alpha|PAXI|Paired box protein Pax-1
- Amino acid number:305 to 559
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:C-terminal part of recombinant chicken paxillin (amino acids 305-559).
- Size:100 µg
- Pk:0,1 mg
Mouse IgG monoclonal antibody for Paxillin, paxillin (PXN) detection. Tested with WB, ICC in Human; mouse; rat; chicken. No cross reactivity with other proteins.
Reconstitute with 1,2% sodium acetate or neutral PBS.
If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.
Type: Primary
Antigen: Paxillin
Clonality: Monoclonal
Clone: PAX-14
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Chicken