Anti-VABP Mouse Monoclonal Antibody [clone: 6C239.4]
Catalog # NOVUNBP2-24876
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Kv channel interacting protein 1
- Antigen symbol:KCNIP1
- Clonality:Monoclonal
- Clone:6C239.4
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Gene ID:30820
- Antigen synonyms:VABP|KCHIP1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: VABP
Clonality: monoclonal
Clone: 6C239.4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human