Anti-IKBKG Mouse Monoclonal Antibody (Biotin) [clone: 72C627]
NOVUNBP2-24816
- Antibody type:Primary
- Antigen name:Inhibitor Of Kappa Light Polypeptide Gene Enhancer in B-cells, kinase gamma
- Antigen symbol:IKBKG
- Clonality:Monoclonal
- Clone:72C627
- Conjugation:Biotin
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Environmentally Preferable:
- Gene ID:8517
- Antigen synonyms:NEMO|IPD2|IMD33|FIP3|IKK-gamma|Fip3p|ZC2HC9|IP|AMCBX1|IP1|FIP-3|IP2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: IKBKG (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma)
Clonality: monoclonal
Clone: 72C627
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse