Anti-GBP1 Rabbit Polyclonal Antibody
NOVUNBP2-03972
- Antibody type:Primary
- Antigen name:Guanylate binding protein 1, interferon-inducible
- Antigen symbol:GBP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:2633
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
The GBP1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to GBP1. This antibody reacts with human, mouse. The GBP1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 5 µg/ml
Type: Primary
Antigen: GBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse