Anti-TRPV1 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: TRPV1 (transient receptor potential cation channel, subfamily V, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Specifications
- Catalog No:
- NOVUNBP1-97417
- NOVUNBP1-97417SS
- Antigen symbol:
- TRPV1
- TRPV1
- Antigen name:
- Transient receptor potential cation channel subfamily V member 1
- Transient receptor potential cation channel subfamily V member 1
- Antigen synonyms:
- TrpV1|VR1|TRPV1|Transient receptor potential cation channel subfamily V member 1|Capsaicin receptor
- TrpV1|VR1|TRPV1|Transient receptor potential cation channel subfamily V member 1|Capsaicin receptor
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 7442
- 7442
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice