Anti-SUV420H1 Rabbit Polyclonal Antibody
The SUV420h1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SUV420h1. This antibody reacts with human, mouse. The SUV420h1 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Western Blot (Negative).
Recommended Dilutions: Immunohistochemistry (Parafin): 1:200
Type: Primary
Antigen: SUV420h1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
- Catalog No:
- NOVUNBP1-97312SS
- NOVUNBP1-97312
- Antigen symbol:
- SUV420H1
- SUV420H1
- Antigen name:
- Suppressor of Variegation 4-20 Homolog 1
- Suppressor of Variegation 4-20 Homolog 1
- Antigen synonyms:
- CGI85|CGI-85|KMT5B
- CGI85|CGI-85|KMT5B
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 51111
- 51111
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice