Order Entry
Sweden
ContactUsLinkComponent
Anti-PIEZO2 Rabbit Polyclonal Antibody
Anti-PIEZO2 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
undefined
Anti-PIEZO2 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNBP1-78624SS
  • NOVUNBP1-78624
  • Antigen symbol:
  • PIEZO2
  • PIEZO2
  • Antigen name:
  • Piezo-type Mechanosensitive Ion Channel Component 2
  • Piezo-type Mechanosensitive Ion Channel Component 2
  • Antigen synonyms:
  • HsT771|C18orf58|FLJ23144|DA5|HsT748|C18orf30|FAM38B|FLJ23403|FAM38B2
  • HsT771|C18orf58|FLJ23144|DA5|HsT748|C18orf30|FAM38B|FLJ23403|FAM38B2
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 63895
  • 63895
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information