Anti-PIEZO1 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
The PIEZO1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PIEZO1. This antibody reacts with human, mouse. The PIEZO1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:400
Type: Primary
Antigen: PIEZO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- NOVUNBP1-78537-A
- NOVUNBP1-78537-B
- Antigen symbol:
- PIEZO1
- PIEZO1
- Antigen name:
- Piezo-type Mechanosensitive Ion Channel Component 1
- Piezo-type Mechanosensitive Ion Channel Component 1
- Antigen synonyms:
- Mib|FAM38A|DHS
- Mib|FAM38A|DHS
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 9780
- 9780
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice