Anti-ABCG8 Mouse Monoclonal Antibody [clone: 1B10A5]
Supplier: Novus Biologicals
Certificates
About this item
The ABCG8 Antibody (1B10A5) from Novus Biologicals is a mouse monoclonal antibody to ABCG8. This antibody reacts with human, mouse. The ABCG8 Antibody (1B10A5) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:100
Type: Primary
Antigen: ABCG8
Clonality: Monoclonal
Clone: 1B10A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- NOVUNBP1-71706SS
- NOVUNBP1-71706
- Antigen symbol:
- ABCG8
- ABCG8
- Antigen name:
- ATP Binding Cassette, sub-family G, member 8
- ATP Binding Cassette, sub-family G, member 8
- Antigen synonyms:
- STSL|ATP-binding cassette sub-family G member 8|Sterolin-2|ABCG8|GBD4
- STSL|ATP-binding cassette sub-family G member 8|Sterolin-2|ABCG8|GBD4
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 1B10A5
- 1B10A5
- Reactivity:
- Mouse
- Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 64241
- 64241
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice