Anti-GPR155 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Katalog # NOVUNBP1-50625F
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:G Protein-Coupled Receptor 155
- Antigen symbol:GPR155
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:151556
- Antigen synonyms:PGR22|DEPDC3|DEP.7
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: GPR155 (G protein-coupled receptor 155)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human