Anti-DUX4 Mouse Monoclonal Antibody (DyLight® 550) [clone: P4H2]
Catalog # NOVUNBP1-49552R
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Double homeobox 4
- Antigen symbol:DUX4
- Clonality:Monoclonal
- Clone:P4H2
- Conjugation:DyLight® 550
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Gene ID:22947
- Antigen synonyms:double homeobox protein 10|DUX10|double homeobox protein 4/10|double homeobox protein DUX10|double homeobox protein 4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: DUX4 (double homeobox 4)
Clonality: monoclonal
Clone: P4H2
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human