Anti-ACTB Mouse Monoclonal Antibody [clone: 8H10D10]
Supplier: Novus Biologicals
Certificates
About this item
The beta-Actin Antibody (8H10D10) from Novus Biologicals is a mouse monoclonal antibody to beta-Actin. This antibody reacts with human, mouse, rat, chinese hamster, primate. The beta-Actin Antibody (8H10D10) has been validated for the following applications: Western Blot, Simple Western, Flow Cytometry, ELISA, Immunocytochemistry / Immunofluorescence.
Simple Western
Type: Primary
Antigen: beta-Actin
Clonality: Monoclonal
Clone: 8H10D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat, Chinese Hamster, Primate
Specifications
- Catalog No:
- NOVUNBP1-47423
- NOVUNBP1-47423SS
- Antigen symbol:
- ACTB
- ACTB
- Antigen name:
- Actin beta
- actin, beta
- Antigen synonyms:
- PS1TP5-binding protein 1|cytoplasmic 1|PS1TP5BP1|BRWS1|beta cytoskeletal actin|actin
- PS1TP5BP1|BRWS1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 8H10D10
- 8H10D10
- Reactivity:
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 60
- 60
- Isotype:
- IgG2b
- IgG2b
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice