Anti-RCAN1 Rabbit Polyclonal Antibody
The RCAN1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to RCAN1. This antibody reacts with human, mouse. The RCAN1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:1000
Type: Primary
Antigen: RCAN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
- Catalog No:
- NOVUNBP1-46852
- NOVUNBP1-46852SS
- Antigen symbol:
- RCAN1
- RCAN1
- Antigen name:
- Regulator of calcineurin 1
- Regulator of calcineurin 1
- Antigen synonyms:
- ADAPT78|DSCR1|Regulator of calcineurin 1|RCAN1|DSC1|Calcipressin-1|Myocyte-enriched calcineurin-interacting protein 1|Adapt78|CSP1|Down syndrome critical region protein 1
- ADAPT78|DSCR1|Regulator of calcineurin 1|RCAN1|DSC1|Calcipressin-1|Myocyte-enriched calcineurin-interacting protein 1|Adapt78|CSP1|Down syndrome critical region protein 1
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 1827
- 1827
- Western blot:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice