Anti-SULF2 Mouse Monoclonal Antibody (DyLight® 550) [clone: 2B4]
NOVUNBP1-36727R
- Antibody type:Primary
- Antigen name:Sulfatase 2
- Clonality:Monoclonal
- Clone:2B4
- Conjugation:DyLight® 550
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Gene ID:55959
- Antigen synonyms:KIAA1247|FLJ44488|MGC126411|HSULF-2|DKFZp313E091|RP5-1049G16.1|FLJ90554
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
The Sulfatase-2 / SULF2 Antibody (2B4) [DyLight 550] from Novus Biologicals is a mouse monoclonal antibody to Sulfatase-2 / SULF2. This antibody reacts with human, mouse. The Sulfatase-2 / SULF2 Antibody (2B4) [DyLight 550] has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:250 - 1:500
Type: Primary
Antigen: SULF2
Clonality: Monoclonal
Clone: 2B4
Conjugation: DyLight 550
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse