Anti-BIRC3 Rabbit Polyclonal Antibody (DyLight® 550)
NOVUNBP1-27972R
- Antibody type:Primary
- Antigen name:baculoviral IAP repeat containing 3
- Antigen symbol:cIAP2
- Clonality:Polyclonal
- Conjugation:DyLight® 550
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:330
- Antigen synonyms:MIHC|MALT2|HAIP1|API2|AIP1|RNF49|CIAP2|HIAP1|c-IAP2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
The cIAP-2 / HIAP-1 Antibody [DyLight 550] from Novus Biologicals is a rabbit polyclonal antibody to cIAP-2 / HIAP-1. This antibody reacts with human. The cIAP-2 / HIAP-1 Antibody [DyLight 550] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:800-1:3200
Type: Primary
Antigen: cIAP-2/HIAP-1
Clonality: Polyclonal
Clone:
Conjugation: DyLight 550
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human