Anti-Ferroportin 1 Rabbit Polyclonal Antibody
Type: Primary
Antigen: Ferroportin 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
- Catalog No:
- NOVUNBP1-21502
- NOVUNBP1-21502SS
- Antigen symbol:
- FPN1
- FPN1
- Antigen name:
- Ferroportin 1
- Ferroportin 1
- Antigen synonyms:
- HFE4|SLC11A3|IREG1|SLC40A1|MST079|solute carrier family 40 member 1|MSTP079|MTP1|FPN1
- HFE4|SLC11A3|IREG1|SLC40A1|MST079|solute carrier family 40 member 1|MSTP079|MTP1|FPN1
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 30061
- 30061
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice