Order Entry
Sweden
ContactUsLinkComponent
Anti-HPi1 Mouse Monoclonal Antibody [clone: HIC0-4F9]
Anti-HPi1 Mouse Monoclonal Antibody [clone: HIC0-4F9]
Supplier:  Novus Biologicals
Certificates
undefined
Anti-HPi1 Mouse Monoclonal Antibody [clone: HIC0-4F9]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNBP1-18872
  • NOVUNBP1-18872SS
  • Antigen symbol:
  • HPI1
  • HPI1
  • Antigen name:
  • Heterochromatin Protein 1
  • Heterochromatin Protein 1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • HIC0-4F9
  • HIC0-4F9
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information