Anti-SIGLEC1 Mouse Monoclonal Antibody [clone: HSn 7D2]
Type: Primary
Antigen: SIGLEC1 (sialic acid binding Ig-like lectin 1, sialoadhesin)
Clonality: monoclonal
Clone: HSn 7D2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
- Catalog No:
- NOVUNB600-534SS
- NOVUNB600-534
- Antigen symbol:
- SIGLEC1
- SIGLEC1
- Antigen name:
- Sialic acid binding Ig-like lectin 1, sialoadhesin
- Sialic acid binding Ig-like lectin 1, sialoadhesin
- Antigen synonyms:
- dJ1009E24.1|SN|CD169|SIGLEC-1
- SIGLEC1|Sialic acid-binding Ig-like lectin 1|dJ1009E24.1|SN|CD169|Sialoadhesin|SIGLEC-1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- HSn 7D2
- HSn 7D2
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 6614
- 6614
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice