Anti-Blimp-1 Mouse Monoclonal Antibody (DyLight® 488) [clone: 3H2-E8]
NOVUNB600-235G
- Antibody type:Primary
- Antigen name:PR Domain Zinc Finger Protein 1
- Antigen symbol:BLIMP1
- Clonality:Monoclonal
- Clone:3H2-E8
- Conjugation:DyLight® 488
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Gene ID:639
- Antigen synonyms:PRDI-BF1|B-lymphocyte-induced maturation protein 1|PR domain zinc finger protein 1|RP1-134E15.1|BLIMP1|BLIMP-1|PRDI-binding factor-1|beta-interferon gene positive-regulatory domain I binding factor
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
The BLIMP1 / PRDM1 Antibody (3H2-E8) [DyLight 488] from Novus Biologicals is a mouse monoclonal antibody to BLIMP1 / PRDM1. This antibody reacts with human, mouse, porcine. The BLIMP1 / PRDM1 Antibody (3H2-E8) [DyLight 488] has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:200
Type: Primary
Antigen: BLIMP1/PRDM1
Clonality: Monoclonal
Clone: 3H2-E8
Conjugation: DyLight 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Porcine