Anti-IL23R Rabbit Polyclonal Antibody
Katalog # NOVUNB600-1147
Leverantör: Novus Biologicals
Specifikationer
- Antibody type:Primary
- Antigen name:Interleukin 23 Receptor
- Antigen symbol:IL23R
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:149233
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 μL
- Pk:0,2 mL
Specifikationer
Om denna produkt
Type: Primary
Antigen: IL23R (interleukin 23 receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse